missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Antibody (R29.6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £342.00
Specifications
| Antigen | Maltose Binding Protein |
|---|---|
| Clone | R29.6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot : 1:100 - 1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694809
|
Novus Biologicals
NBP2-76651-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18384851
|
Novus Biologicals
NBP2-76651 |
0.1 mL |
£342.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Maltose Binding Protein | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| PBS with 0.02% sodium azide | |
| MOS maltose binding protein fusion protein | |
| Primary | |
| This antibody is useful for detection and isolation of recombinant MBP fusion proteins. |
| R29.6 | |
| Western Blot : 1:100 - 1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Unconjugated | |
| Non-species specific | |
| Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title