missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Maltose Binding Protein Polyclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Maltose Binding Protein |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000 |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide |
| Gene Alias | Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein |
| Host Species | Rabbit |
| Immunogen | This antibody was purified from whole rabbit serum prepared by repeated immunizations with the Maltose Binding Protein epitope tag recombinant protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?