missing translation for 'onlineSavingsMsg'
Learn More

Maltol, MedChemExpress

Product Code. 16409366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16409366 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 16409366

Brand: Medchem Express HYW012788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maltol, a type of aromatic compound, exists in high concentrations in red ginseng. Maltol is a potent antioxidative agent and typically is used to enhance flavor and preserve food.

Maltol, a type of aromatic compound, exists in high concentrations in red ginseng. Maltol is a potent antioxidative agent and typically is used to enhance flavor and preserve food.

Specifications

Chemical Name or Material Maltol
CAS 118-71-8
Color White
Purity Grade Notes Research
Molecular Formula C6H6O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (792.96 mM; Need ultrasonic) ∣H2O : 1.82 mg/mL (14.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C(O)=C(C)OC=C1
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.