missing translation for 'onlineSavingsMsg'
Learn More

Maltitol (D-Maltitol), Mikromol™

Product Code. 17927804
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17927804 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17927804 Supplier Mikromol Supplier No. MM3570.00
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Maltitol (D-Maltitol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maltitol
CAS 585-88-6
Analyte or Component Names Maltitol
Shipping Condition Room Temperature
Molecular Formula C12 H24 O11
Certifications/Compliance ISO 17034
Synonym Maltitol (6CI,7CI), Amalty, Amalty MR, Amalty MR 100, Amalty MR 20, Amalty MR 50, Amalty P, Amalty Syrup, Cerestar 16303, D-Maltitol, Lesys, Lycasin HBC, Mabit, Mabit 50M, Malbit CH, Malbit CH 16385, Malbit CR, Malti MR, Maltidex, Maltidex 100, Maltidex CH 16385, Maltidex H 16330, Maltidex M 16313, Maltilite 5575, Maltilite 7575, Maltilite P 200, Maltisorb, Maltisorb 75/77, Maltisorb P 200, Maltisorb P 35, Maltisorb P 90, Maltisweet, Maltisweet 3145, Maltit, Maltitol G 3, Sweet G 2, SweetPearl, SweetPearl P 200, SweetPearl P 35, Glucitol, 4-O-alpha-D-glucopyranosyl-, D- (8CI), 4-O-alpha-D-Glucopyranosyl-D-glucitol, Maltilol, GR 155535X, Erythritol Impurity A, Mannitol Impurity B, Sorbitol Impurity C
InChI Formula InChI=1S/C12H24O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h4-21H,1-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+/m0/s1
SMILES OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)CO
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
Molecular Weight (g/mol) 344.31
Quantity 250 mg
Formula Weight 344.1319 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.