Learn More
Invitrogen™ MALT1 Recombinant Rabbit Monoclonal Antibody (24GB440)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557058
 View more versions of this product
Product Code. 30276521
  • £126.00 / 20µL
Description
MALT1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MALT1
Recombinant Monoclonal
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UDY8
MALT1
A synthesized peptide derived from human MALT1 (aa 1-60).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB440
Unconjugated
MALT1
A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1
Rabbit
Affinity Chromatography
RUO
10892
-20°C