missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MALT1 Recombinant Rabbit Monoclonal Antibody (24GB440)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557057
This item is not returnable.
View return policy
Description
MALT1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
Specifications
| MALT1 | |
| Recombinant Monoclonal | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UDY8 | |
| MALT1 | |
| A synthesized peptide derived from human MALT1 (aa 1-60). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB440 | |
| Unconjugated | |
| MALT1 | |
| A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10892 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction