missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517358
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
Specifications
| MALT1 | |
| Polyclonal | |
| Unconjugated | |
| MALT1 | |
| A630046N12; caspase-like protein; D430033E09Rik; DKFZp434L132; IMD12; MALT lymphoma-associated translocation; Malt1; malt-1; MALT1 paracaspase; MALT1 protease; MLT; MLT1; mucosa associated lymphoid tissue lymphoma translocation gene 1; mucosa-associated lymphoid tissue lymphoma translocation protein 1; Mucosa-associated lymphoid tissue lymphoma translocation protein 1 homolog; paracaspase; paracaspase 1; paracaspase-1; PCASP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10892, 240354, 307366 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 29 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
| Q2TBA3, Q9UDY8 | |
| MALT1 | |
| Synthetic peptide corresponding to the carboxy terminus of mouse and rat MALT1 protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction