missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MALT1 Mouse anti-Human, Clone: 4D3F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66225-1-IG-150UL
This item is not returnable.
View return policy
Description
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma. MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.Specifications
| MALT1 | |
| Monoclonal | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UDY8 | |
| MALT1 | |
| MALT1 Fusion Protein Ag20632 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 4D3F6 | |
| Unconjugated | |
| MALT1 | |
| MALT1, MLT, MLT1, Paracaspase | |
| Mouse | |
| Protein G | |
| RUO | |
| 10892 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction