missing translation for 'onlineSavingsMsg'
Learn More

MALT1 inhibitor MI-2, MedChemExpress

Product Code. 16481287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481287 50 mg 50mg
16447777 5 mg 5mg
16425796 100 mg 100mg
16405058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481287 Supplier Medchem Express Supplier No. HY12276
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MALT1 inhibitor MI-2 is a MALT1 inhibitor (IC50=5.84 μM). MALT1 inhibitor MI-2 binds directly to MALT1, irreversibly suppresses protease function and is accompanied by NF-κB reporter activity suppression, c-REL nuclear localization inhibition, and NF-κB target gene downregulation. MALT1 inhibitor MI-2 shows nontoxic to animals.

MALT1 inhibitor MI-2 is a MALT1 inhibitor (IC50=5.84 μM). MALT1 inhibitor MI-2 binds directly to MALT1, irreversibly suppresses protease function and is accompanied by NF-κB reporter activity suppression, c-REL nuclear localization inhibition, and NF-κB target gene downregulation. MALT1 inhibitor MI-2 shows nontoxic to animals.

Specifications

Chemical Name or Material MALT1 inhibitor MI-2
CAS 1047953-91-2
Color White
Purity Grade Notes Research
Molecular Formula C19H17Cl3N4O3
Quantity 50 mg
Solubility Information DMSO : ≥ 46 mg/mL (100.94 mM)
SMILES O=C(NC1=CC=C(N2N=C(OCCOC)N=C2C3=CC=C(Cl)C(Cl)=C3)C=C1)CCl
Molecular Weight (g/mol) 455.72
Formula Weight 455.72
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.