missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MALT1 Monoclonal specifically detects MALT1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MALT1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rMT1/410 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin : 1-2 ug/mL |
| Formulation | 10mM PBS |
| Gene Alias | caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?