missing translation for 'onlineSavingsMsg'
Learn More

Malt Extract, MP Biomedicals

Product Code. 11427582 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g

Product Code. 11427582

Brand: MP Biomedicals 0215531580

Please to purchase this item. Need a web account? Register with us today!

From starch digestion. A source of carbohydrates

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-48-0

Specifications

Quantity 100 g
Chemical Name or Material Malt Extract
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.