missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malotilate, MedChemExpress
Description
Malotilate (NKK 105), an orally active hepatotropic agent and an anti-fibrotic substance, selectively inhibits the 5-lipoxygenase (5-LOX) (IC50=4.7 μM). Malotilate prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels and preventing the retention of transferrin in the hepatocytes.
Specifications
Specifications
| Chemical Name or Material | Malotilate |
| CAS | 59937-28-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C12H16O4S2 |
| Quantity | 50 mg |
| Synonym | NKK 105 |
| Solubility Information | DMSO : ≥ 100 mg/mL (346.76 mM) |
| SMILES | O=C(OC(C)C)/C(C(OC(C)C)=O)=C1SC=CS\1 |
| Molecular Weight (g/mol) | 288.38 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?