missing translation for 'onlineSavingsMsg'
Learn More

Malonyl dichloride, 94%

Product Code. 15496298
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496298 100 g 100g
15486298 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496298 Supplier Thermo Scientific Alfa Aesar Supplier No. H59868.22
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1663-67-8 | C3H2Cl2O2 | 140.947 g/mol

Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonyl Dichloride is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Decomposes in water.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-67-8
Molecular Formula C3H2Cl2O2
Molecular Weight (g/mol) 140.947
MDL Number MFCD00000735
InChI Key SXYFKXOFMCIXQW-UHFFFAOYSA-N
Synonym malonyl chloride, malonyl dichloride, malonoyl chloride, malonic acid dichloride, malonoyl dichloride, malonic acid chloride, malonylchloride, methane-1,1-dicarbonyl chloride, malonylchlorid, malonyldichloride
PubChem CID 74269
IUPAC Name propanedioyl dichloride
SMILES C(C(=O)Cl)C(=O)Cl

Specifications

Density 1.449
Boiling Point 53°C to 55°C (19 mmHg)
Flash Point 47°C (117°F)
Linear Formula CH2(COCl)2
Refractive Index 1.465
Quantity 100 g
UN Number UN2920
Beilstein 774044
Solubility Information Decomposes in water.
Formula Weight 140.95
Percent Purity 97%
Chemical Name or Material Malonyl dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.