missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, 99%

Product Code. 10200020 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10200020

Brand: Thermo Scientific Acros 125270025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-77-3 | C3H2N2 | 66.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
Molecular Weight (g/mol) 66.06
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile
SMILES C(C#N)C#N

Specifications

Melting Point 30.0°C to 34.0°C
Density 1.0400g/mL
Boiling Point 220.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2CN
Quantity 2.5 kg
Beilstein 02,589
Fieser 04,317
Merck Index 15,5775
Specific Gravity 1.04
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Formula Weight 66.06
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Malononitrile, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Malononitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.