missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid disodium salt hydrate, 99%

Product Code. 10645501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10645501 25 g Plastic bottle 25g
10685681 100 g Plastic bottle 100g
10165503 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10645501 Supplier Thermo Scientific Acros Supplier No. 413500250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132499-82-2 | C3H2Na2O4 | 148.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132499-82-2
Molecular Formula C3H2Na2O4
Molecular Weight (g/mol) 148.03
MDL Number MFCD00149114
InChI Key PRWXGRGLHYDWPS-UHFFFAOYSA-L
IUPAC Name disodium propanedioate
SMILES [Na+].[Na+].[O-]C(=O)CC([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH2(COONa)2·xH2O
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 148.03
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Malonic acid disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.