missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid, 99%, MP Biomedicals™

Product Code. 11349109 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349109 1 kg 1kg
11329109 100 g 100g
11339109 250 g 250g
11462440 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11349109 Supplier MP Biomedicals Supplier No. 0210223891
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.061
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O

Specifications

Color White
Quantity 1 kg
Formula Weight 104.1
Percent Purity 0.99
Physical Form Powder
Chemical Name or Material Malonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.