missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid-1,2,3-13C3, TRC

Product Code. 30483827
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483827 10 mg 10mg
30481824 100 mg 100mg
30470208 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483827 Supplier Toronto Research Chemicals Supplier No. TRCM15800910MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102342-85-8
Molecular Formula 13C3H4O4
Molecular Weight (g/mol) 107.04
Synonym Propanedioic Acid-1,2,3-13C3, 1,3-Propanedioic Acid-1,2,3-13C3, Carboxyacetic Acid-1,2,3-13C3, Dicarboxymethane-1,2,3-13C3, Methanedicarboxylic Acid-1,2,3-13C3, NSC 8124-2-1,2,3-13C3, [1,2,3-13C3]-Malonic acid
SMILES O[13C]([13C]([H])([H])[13C](O)=O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)/i1+1,2+1,3+1
Chemical Name or Material Malonic Acid-1,2,3-13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.