missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Malondialdehyde Monoclonal antibody specifically detects Malondialdehyde in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
Specifications
| Antigen | Malondialdehyde |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 6H6 |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 3-Propanedial;Malonaldehyde;Malondialdehyde;MDA |
| Host Species | Mouse |
| Immunogen | Synthetic Malondialdehyde modified Keyhole Limpet Hemocyanin (KLH). |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?