missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(diethyl acetal), 97%

Product Code. 10295720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295720 5 mL Glass bottle 5mL
10317620 100 mL Glass bottle 100mL
10560204 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295720 Supplier Thermo Scientific Acros Supplier No. 156730050
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 122-31-6 | C11H24O4 | 220.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-31-6
Molecular Formula C11H24O4
Molecular Weight (g/mol) 220.31
MDL Number MFCD00009240
InChI Key KVJHGPAAOUGYJX-UHFFFAOYSA-N
Synonym malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal
PubChem CID 67147
IUPAC Name 1,1,3,3-tetraethoxypropane
SMILES CCOC(CC(OCC)OCC)OCC

Specifications

Melting Point -90.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 220.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C2H5O)2CHCH2CH(OC2H5)2
Refractive Index 1.4100 to 1.4120
Quantity 5 mL
Beilstein 01, III, 3095
Fieser 02,257
Specific Gravity 0.91
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Formula Weight 220.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Malonaldehyde bis(diethyl acetal)
Show More Show Less
compliance-icons
Product Identifier
  • Malonaldehyde bis(diethyl acetal)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.