missing translation for 'onlineSavingsMsg'
Learn More

Malic Acid, Mikromol™

Product Code. 17982126
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982126 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17982126 Supplier Mikromol Supplier No. MM1332.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Malic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Malic Acid
CAS 6915-15-7
Analyte or Component Names DL-Malic Acid
Shipping Condition Room Temperature
Molecular Formula C4 H6 O5
Certifications/Compliance ISO 17034
Synonym Malic acid, (2RS)-2-Hydroxybutanedioic acid, Butanedioic acid, hydroxy- (9CI), Malic acid (8CI), 2-Hydroxybutanedioic acid, 2-Hydroxyethane-1,2-dicarboxylic acid, 2-Hydroxysuccinic acid, DL-Malic acid, Deoxytetraric acid, E 296, FDA 2018, Hydroxybutanedioic acid, Hydroxysuccinic acid, Musashi-no-Ringosan, NSC 25941, Nanoveson M, Pomalus Acid, Xeros, dl-Malic acid, (+/-)-Malic acid, R,S(+/-)-Malic acid, alpha-Hydroxysuccinic acid, Aspartic Acid Impurity A, Magnesium Aspartate Dihydrate Impurity B, Magnesium Aspartate Impurity B
InChI Formula InChI=1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)
SMILES OC(CC(=O)O)C(=O)O
IUPAC Name 2-hydroxybutanedioic acid
Molecular Weight (g/mol) 134.09
Quantity 250 mg
Formula Weight 134.0215 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.