missing translation for 'onlineSavingsMsg'
Learn More

Maleimide, TRC

Product Code. 30482686
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
Unit Size:
10g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482686 10 g 10g
30473972 2 g 2g
30481827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482686 Supplier Toronto Research Chemicals Supplier No. TRCM12498010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-59-3
Molecular Formula C4H3NO2
Molecular Weight (g/mol) 97.07
Synonym 2,5-Dioxo-2,5-dihydropyrrole, 3-Pyrroline-2,5-dione, Maleic Imide, NSC 13684
IUPAC Name pyrrole-2,5-dione
SMILES O=C1NC(=O)C=C1

Specifikationer

Quantity 10 g
InChI Formula InChI=1S/C4H3NO2/c6-3-1-2-4(7)5-3/h1-2H,(H,5,6,7)
Formula Weight 97.0164
Chemical Name or Material Maleimide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.