missing translation for 'onlineSavingsMsg'
Learn More

Maleimide, 98+%

Product Code. 11424504
Click to view available options
Quantity:
5 g
25 g
50 g
250 g
Unit Size:
250g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11424504

Brand: Thermo Scientific Alfa Aesar A13135.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-59-3 | C8H4AgKN2O4 | 339.10 g/mol

Reacts quantitatively with sulfhydryl groups.Maleimide is used to immobilize the bovine serum albumin-boronic acid conjugates in association with silica beads. Its nanoparticle surface favors the conjugation with cell penetration peptides. It is involved in Rhodium-catalyzed conjugate arylation with arylboronic acids. It is actively involved in Michael additions and Diels-Alder reactions by the addition across the double bond. It acts as crosslinking reagents in polymer chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reacts quantitatively with sulfhydryl groups.Maleimide is used to immobilize the bovine serum albumin-boronic acid conjugates in association with silica beads. Its nanoparticle surface favors the conjugation with cell penetration peptides. It is involved in Rhodium-catalyzed conjugate arylation with arylboronic acids. It is actively involved in Michael additions and Diels-Alder reactions by the addition across the double bond. It acts as crosslinking reagents in polymer chemistry.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-59-3
Molecular Formula C8H4AgKN2O4
Molecular Weight (g/mol) 339.10
MDL Number MFCD00005494
InChI Key KWLWLQRUFFGWNG-UHFFFAOYSA-L
Synonym maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one
PubChem CID 10935
ChEBI CHEBI:16072
IUPAC Name pyrrole-2,5-dione
SMILES [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1

Specifications

Melting Point 90°C to 94°C
Density 1.249
Boiling Point 97°C to 103°C (5 mmHg)
Quantity 50 g
UN Number UN2923
Beilstein 106910
Solubility Information Soluble in water.
Formula Weight 97.07
Percent Purity ≥98%
Chemical Name or Material Maleimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.