missing translation for 'onlineSavingsMsg'
Learn More

Maleic Hydrazide, TRC

Product Code. 30484198
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484198 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30484198 Supplier Toronto Research Chemicals Supplier No. TRCM124500100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic hydrazide
CAS 123-33-1
Molecular Formula C4 H4 N2 O2
Quantity 100 g
Synonym 3,6-Pyridazinedione, 1,2-dihydro-, 3(2H)-Pyridazinone, 6-hydroxy- (6CI,7CI), 1,2-Dihydro-3,6-pyridazinedione, 1,2,3,6-Tetrahydropyridazine-3,6-dione, 3,6-Dihydroxypyridazine, 3,6-Dioxopyridazine, 3,6-Pyridazinediol, 6-Hydroxy-3(2H)-pyridazinone, Antergon, Antyrost, MAH, MH, MH (hydrazide), Maleic acid cyclic hydrazide, Maleic cyclic hydrazide, Maleic hydrazide, Malepin, Malzid, Mazide, NSC 13892, NSC 234167, NSC 48832, NSC 8823, Regulox 36, Royal Slo-Gro, Sorbatran, Super De-Sprout, Yadi, Yake
InChI Formula InChI=1S/C4H4N2O2/c7-3-1-2-4(8)6-5-3/h1-2H,(H,5,7)(H,6,8)
SMILES O=C1NNC(=O)C=C1
IUPAC Name 1,2-dihydropyridazine-3,6-dione
Molecular Weight (g/mol) 112.09
Formula Weight 112.0273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.