missing translation for 'onlineSavingsMsg'
Learn More

Maleic hydrazide, 97%, Thermo Scientific™

Product Code. p-7020540
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11484194

Brand: Thermo Scientific Alfa Aesar A12531.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Maleic hydrazide is a synthetic plant growth regulator that is widely used for controlling growth and to induce dormancy in some crops.

Solubility
Soluble in water (4.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-33-1
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00006665
InChI Key BGRDGMRNKXEXQD-UHFFFAOYSA-N
Synonym pyridazine-3,6-diol, maleic hydrazide, 3,6-dihydroxypyridazine, 3,6-pyridazinediol, stuntman, maleic acid hydrazide, vondalhyde, 3,6-pyridazinedione, 1,2-dihydro, 1,2-dihydro-3,6-pyridazinedione, antergon
PubChem CID 21954
ChEBI CHEBI:81771
IUPAC Name 1,2-dihydropyridazine-3,6-dione
SMILES C1=CC(=O)NNC1=O

Specifications

Melting Point ∼305°C (decomposition)
Density 1.6
Flash Point 300°C (572°F)
Quantity 100 g
Beilstein 112223
Merck Index 14,5705
Solubility Information Soluble in water (4.5g/L).
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material Maleic hydrazide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.