missing translation for 'onlineSavingsMsg'
Learn More

Maleic Anhydride, TRC

Product Code. 30483059
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30483059

Brand: Toronto Research Chemicals TRCM124400100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic anhydride
CAS 108-31-6
Molecular Formula C4 H2 O3
Quantity 100 g
Synonym 2,5-Furandione, Maleic anhydride (8CI), 2,5-Dihydrofuran-2,5-dione, A 1018, BM 10, Baolilong A 1018, Crystal Man, Crystal Man AB, Dihydro-2,5-dioxofuran, GR 216, HA 59, HG 3-993-76, M 116389, M 188, Maleic acid anhydride, NSC 137651, NSC 137652, NSC 137653, NSC 9568, Nourymix MA 901, SY-A, Toxilic anhydride, WS 8201, cis-Butenedioic anhydride
InChI Formula InChI=1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H
SMILES O=C1OC(=O)C=C1
IUPAC Name furan-2,5-dione
Molecular Weight (g/mol) 98.06
Formula Weight 98.0004
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.