missing translation for 'onlineSavingsMsg'
Learn More

Maleic anhydride, briquettes

Product Code. 16554324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
Unit Size:
1kg
25g
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554324 1 kg 1kg
16574324 25 g 25g
16564324 5 kg 5kg
16584324 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16554324 Supplier Thermo Scientific Acros Supplier No. 465660010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-31-6 | C4H2O3 | 98.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-31-6
Molecular Formula C4H2O3
Molecular Weight (g/mol) 98.06
MDL Number MFCD00005518
InChI Key FPYJFEHAWHCUMM-UHFFFAOYSA-N
IUPAC Name 2,5-dihydrofuran-2,5-dione
SMILES O=C1OC(=O)C=C1

Specifications

Melting Point 52.0°C to 55.0°C
Boiling Point 200.0°C
Flash Point 102°C
Quantity 1 kg
Solubility Information Solubility aq. soln.: 400 g/L (25°C). Other solubilities: reacts with water forming maleic acid, 227 g/100 g acetone - 50 g/100 g benzene (25°C), 112 g/100 g ethylacetate (25°C)- 0,60 g/100 g ccl4, 52.5 g/100 g chloroform (25°C)-23,4 g/100 g toluen
Viscosity 1.6 mPa.s (60°C)
Formula Weight 98.06
Chemical Name or Material Maleic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.