missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, TRC

Product Code. 30473093
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473093 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473093 Supplier Toronto Research Chemicals Supplier No. TRCM124300250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maleic Acid
CAS 110-16-7
Purity Grade Notes HPLC
Molecular Formula C4 H4 O4
Quantity 250 g
Synonym (Z)-2-Butenedioic acid, Maleic acid, (2Z)-Butene-2-dioic acid, (Z)-2-Butenedioic acid, Maleinic acid, Malezid CM, Scotchbond Multipurpose Etchant, Toxilic acid, cis-1,2-Ethylenedicarboxylic acid, cis-2-Butenedioic acid, cis-Butenedioic acid
InChI Formula InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-
SMILES OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
Formula Weight 116.011
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.