missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, MedChemExpress

Product Code. 16472708
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472708 Supplier Medchem Express Supplier No. HYY0367
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E. coli and L. monocytogenes.

Maleic Acid is a Glutamate Decarboxylase (GAD) inhibitor of E. coli and L. monocytogenes.

Specifications

Chemical Name or Material Maleic Acid
CAS 110-16-7
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C4H4O4
Quantity 5 g
Solubility Information H2O : 100 mg/mL (861.55 mM; Need ultrasonic)
SMILES O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 116.07
Formula Weight 116.07
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.