missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, Certified AR for Analysis, Fisher Chemical

Product Code. 10598430 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10598430 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10598430 Supplier Fisher Chemical Supplier No. M/1255/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 110-16-7
Melting Point 134°C
Color White
pH 1.3
Packaging Plastic powder jar
Molecular Formula C4H4O4
MDL Number MFCD00063177
Quantity 500 g
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES OC(=O)\C=C\C(O)=O
IUPAC Name (Z)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
PubChem CID 444266
ChEBI CHEBI:18300
Formula Weight 116.07g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.