missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, 99%

Product Code. 10033853 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10033853 25 kg Fibre drum 25kg
10396760 100 g Plastic bottle 100g
10439050 1 kg Plastic bottle 1kg
10348843 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10033853 Supplier Thermo Scientific Acros Supplier No. 125230250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (Z)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 134.0°C to 138.0°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH=CHCO2H
Beilstein 02, IV, 2199
Merck Index 15, 5767
Solubility Information Solubility in water: 790g/L (25°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone, slightly soluble in ether and benzene, practically insoluble in carbon tetrachloride
Formula Weight 116.07
Percent Purity 99%
Quantity 25 kg
Flash Point 127°C
Infrared Spectrum Authentic
Packaging Fibre drum
Physical Form Crystalline Powder
Chemical Name or Material Maleic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.