missing translation for 'onlineSavingsMsg'
Learn More

Maleic acid, 98+%

Product Code. p-7022020
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492018 250 g 250g
11482018 1000 g 1000g
11402028 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11492018 Supplier Thermo Scientific Alfa Aesar Supplier No. A14596.30
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 110-16-7 | C4H4O4 | 116.07 g/mol

Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

Solubility
Soluble in water, acetone, glacial acetic acid, alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.

Notes
Incompatible with oxidizing agents, bases, reducing agents, metals and alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (Z)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 132°C to 136°C
Beilstein 605762
Merck Index 14,5703
Solubility Information Soluble in water,acetone,glacial acetic acid,alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.
Formula Weight 116.07
Percent Purity ≥98%
Density 1.59
Quantity 250 g
Flash Point 127°C (260°F)
Chemical Name or Material Maleic acid
compliance-icons compliance-icons
Product Identifier
  • Maleic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Maleic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.