missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malectin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26655-1-AP-20UL
This item is not returnable.
View return policy
Description
Carbohydrate-binding protein with a strong ligand preference for Glc2-N-glycan. May play a role in the early steps of protein N-glycosylation.Specifications
| Malectin | |
| Polyclonal | |
| Unconjugated | |
| MLEC | |
| KIAA0152, malectin, MLEC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109154, 304543, 9761 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14165, Q5FVQ4, Q6ZQI3 | |
| MLEC | |
| Malectin Fusion Protein Ag24379 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction