Learn More
Value Plastics Male Luer Slip to Female Luer Thread Adapter Fitting
Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Polycarbonate
Polypropylene
missing translation for 'unitSize'
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Inlet | Male Luer Slip, Female Luer Thread |
| Material (Seal) | Polycarbonate |