Learn More
Value Plastics Male Luer Integral Lock Ring to UNF Thread Adapters
Product Code. 30054353 Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
Product Code. 30054353
Brand: Value Plastics 5012087
Only null left
Add to basket
Add to basket
Seal luer ports with a secure and leak-proof connection
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Inlet Male Luer Integral Lock Ring, 1/4-28 UNF(M) Thread
Material (Seal) Nylon
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.