Learn More
Value Plastics Male Luer Integral Lock Ring to Plug Adapters
Product Code. 30054073 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Nylon
Polycarbonate
polypropylene
Unit Size:
Pack of 25
Product Code. Material (Seal) unitSize
30054073 Nylon Pack of 25
30054009 Polycarbonate Pack of 25
30053291 polypropylene Pack of 25
3 options
Product Code. 30054073
Brand: Value Plastics 5012074
Only null left
Add to basket
Add to basket
Seal luer ports with a secure and leak-proof connection
Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. Prevent leaks with integral lock ring mechanism
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Inlet Male Luer Integral Lock Ring, Plug, Closed at Luer Tip
Material (Seal) Nylon