Learn More
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Product Code. p-200105103 Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
Product Code. 30053948
Brand: Value Plastics 5012003
Only null left
Add to basket
Add to basket
Prevent leaks with integral lock ring mechanism
Luer fittings allow for a simple yet secure twist-lock connection. Easily transition from a male luer to female luer fitting. Ideal for general laboratory and single-use applications
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Inlet Male Luer Integral Lock Ring, Female Luer Thread
Material (Seal) Nylon
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.