Learn More
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Product Code. 30053831 Shop All Cole-Parmer Products
Change view
Click to view available options
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Seal) unitSize
30053831 Polypropylene Pack of 25
30053948 Nylon Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053831 Supplier Value Plastics Supplier No. 5012004
Only null left
Add to basket
Add to basket
Prevent leaks with integral lock ring mechanism
Luer fittings allow for a simple yet secure twist-lock connection. Easily transition from a male luer to female luer fitting. Ideal for general laboratory and single-use applications
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Inlet Male Luer Integral Lock Ring, Female Luer Thread
Material (Seal) Polypropylene
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.