Learn More
Value Plastics Male Luer with Integral Lock Ring to 500 Series Barb
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
Material:
Nylon
Polypropylene
Unit Size:
Pack of 25
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 1/8 in. |
| Material | Polypropylene |
| Inlet | Male Luer x Hose Barb |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.