Learn More
Value Plastics Male Luer with Integral Lock Ring to 400 Series Barb
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
3/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Diameter (English) Inner | 1/16 in. |
| Material | Polycarbonate |
| Inlet | Male Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.