Learn More
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Product Code. 30053606 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Material unitSize
30053606 3/32 in. Nylon Pack of 25
30053313 1/16 in. Nylon Pack of 25
30053251 3/16 in. Polypropylene Pack of 25
30053139 1/16 in. Polycarbonate Pack of 25
30054036 1/8 in. Polypropylene Pack of 25
30053588 5/32 in. Polycarbonate Pack of 25
30053386 1/8 in. Nylon Pack of 25
30053124 3/32 in. Polypropylene Pack of 25
30053144 1/8 in. Polycarbonate Pack of 25
30053460 3/16 in. Polycarbonate Pack of 25
30053779 1/16 in. Polypropylene Pack of 25
30053050 3/32 in. Polycarbonate Pack of 25
30053359 5/32 in. Polypropylene Pack of 25
30053180 5/32 in. Nylon Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 30053606 Supplier Value Plastics Supplier No. 5012023
  • ONLINE EXCLUSIVE PRICE
    £18.15
    £16.69 / Pack of 25
    Save £1.46
    8% Off
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Material Nylon
Inlet Male Luer x Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.