Learn More
Value Plastics Male Luer to 500 Series Barb
Product Code. p-200105121 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/4 in.
1/8 in.
Unit Size:
Pack of 25
Product Code. 30053872
Brand: Value Plastics 5012012
Only null left
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/8 in.
Material Polypropylene
Inlet Male Luer x Hose Barb
Always test the application to verify the performance of the interconnection.