Learn More
Value Plastics Male Luer to 500 Series Barb
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/4 in.
1/8 in.
Unit Size:
Pack of 25
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 1/8 in. |
| Material | Polypropylene |
| Inlet | Male Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.