missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Malachite Green Stain

Product Code. 12948647
Change view
Click to view available options
Quantity:
500mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12948647 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12948647 Supplier Pro-Lab Diagnostics™ Supplier No. PL.7030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use as a counterstain during Ziehl-Neelsen and Kinyoun staining. Pro-Lab Diagnostics™ Malachite Green Stain is a ready-to-use stain to differentiate non-acid-fast organisms from mycobacterium. It is manufactured to the highest standards, according to all regulatory requirements, and CE labelled.

Pro-Lab Diagnostics™ Microbiological Stain solutions are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all solutions offer excellent performance characteristics.

Acid-fast organisms, such as Mycobacterium, have wax-like, nearly impermeable cell walls that make them highly resistant to typical microbiological stains. They require staining via either the Ziehl-Neelsen (ZN) method, which requires heating, or the Kinyoun method, which does not. The basic method seen in Gram staining is used: a primary stain is applied, a differentiator removes the stain from non-acid-fast organisms and a counterstain is then applied to identify the decolored organisms.

  • Ready-to-use solution in a convenient, economical size
  • Acts as a counter-stain for non-acid-fast organisms in the Ziehl-Neelsen or Kinyoun staining method
  • Apply only after staining with Carbol Fuchsin and using a differentiator/decolorizer
  • Decolorized non-acid-fast cells take up the contrasting green color for easy detection

Specifications

Solution Type Reagent
Type Malachite Green Stain
Quantity 500mL
Certifications/Compliance (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Format Liquid
For Use With (Application) For Mycobacteria acid fast stains
Color Green
Boiling Point 78–100°C at 1013hPa
Composition Ethanol, Methanol, Malachite Green Oxalate
Storage Requirements <20°C
Stain Type Acid Fast Stain
Show More Show Less
compliance-icons
Product Identifier
  • Malachite Green
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, methanol, malachite green oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.