missing translation for 'onlineSavingsMsg'
Learn More

Malachite Green Oxalate, TRC

Product Code. 30040617
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040617 10 g 10g
18023740 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040617 Supplier Toronto Research Chemicals Supplier No. TRCM11040010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-29-8
Molecular Formula 2 C23 H25 N2 . C2 H2 O4 . 2 C2 H O4
Molecular Weight (g/mol) 927
Synonym Methanaminium, N-[4-[[4-(dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, ethanedioate, ethanedioate (2:2:1), Ammonium, [4-[p-(dimethylamino)-α-phenylbenzylidene]-2,5-cyclohexadien-1-ylidene]dimethyl-, oxalate (1:1), oxalate (1:1) (8CI), Malachite green oxalate (6CI), Ethanedioic acid, ion(1-), N-[4-[[4-(dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-Nmethylmethanaminium, ethanedioate (2:1) (9CI), C. I. Basic Green, Malachite green oxalate salt
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccccc3.CN(C)c4ccc(cc4)C(=C5C=CC(=[N+](C)C)C=C5)c6ccccc6.OC(=O)C(=O)O.OC(=O)C(=O)[O-].OC(=O)C(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2
Formula Weight 926.3738
Chemical Name or Material Malachite green oxalate salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.