missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Malachite Green oxalate, pure, high purity biological stain

Product Code. 10573971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573971 500 g Glass bottle 500g
10503211 25 g Glass bottle 25g
10503401 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10573971 Supplier Thermo Scientific Chemicals Supplier No. 229785000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Malachite Green oxalate, C52H54N4O12, CAS Number-2437-29-8, C.I. 42000, Basic Green 4, 500g, 0.9 to 1.15, 13, 745, 219-441-7, 927.03, Pure, Danger | CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym Basic Green 4, C.I. 42000
PubChem CID 2724411
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;2-hydroxy-2-oxoacetate;oxalic acid
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Absorbance 1650 to 1950 (at lambda max.)
Melting Point 164.0°C
Absorption Ratio 0.9 to 1.15
Beilstein 13, 745
Merck Index 15, 5763
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol, methanol, amyl alcohol
Infrared Spectrum Authentic
Lambda Max 620 to 625nm (in 50:50 IMS:water)
Formula Weight 927.03
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 500 g
Physical Form Crystalline Powder or Crystals With Metal Luster
Chemical Name or Material Malachite Green oxalate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Malachite Green oxalate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H361d-Suspected of damaging the unborn child.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.