missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAK16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47309-25ul
This item is not returnable.
View return policy
Description
MAK16 Polyclonal specifically detects MAK16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MAK16 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| MAK16 homolog (S. cerevisiae), MAK16L, NNP78, protein MAK16 homolog, Protein RBM13, RBM13, RNA binding motif protein 13, RNA binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MAK16 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FIRHKCKQRFTKITQYLIRIRKLTLKRQRKLVPLSKKVERREKRREEKALIAAQLDNAIEKELLERLKQDTYGDIYNFPIHAFDKALEQQ | |
| 25 μL | |
| DNA replication Transcription Translation and Splicing | |
| 84549 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction