missing translation for 'onlineSavingsMsg'
Learn More

MAK10 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18406782 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406782 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406782 Supplier Novus Biologicals Supplier No. NBP177056IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAK10 Polyclonal antibody specifically detects MAK10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAK10
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias bA379P1.1, corneal wound healing-related protein, EGAP, embryonic growth-associated protein homolog, FLJ21613, FLJ22643, MAK10, MAK10 homolog, amino-acid N-acetyltransferase subunit, N(alpha)-acetyltransferase 35, NatC auxiliary subunit, N-alpha-acetyltransferase 35, NatC auxiliary subunit, protein MAK10 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MAK10. The immunogen is located within amino acids 190 - 240 of MAK10. Amino Acid Squence: KDVEDDMQRRVKSTRSRQGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60560
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.