missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Major Environmental Elements: Na, Ca, K, Mg @ 10,000 μg/mL in 2% HNO3, VHG™
Description
Major Environmental Elements: Na, Ca, K, Mg @ 10,000 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Calcium/Magnesium/Potassium/Sodium |
| CAS | 7440-70-2, 7439-95-4, 9-7-7440, 7440-23-5 |
| Methods | ICP/ICPMS |
| Analyte or Component Names | Calcium/Magnesium/Potassium/Sodium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 2% HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Calcium 10000 μg/mL/Magnesium 10000 μg/mL/Potassium 10000 μg/mL/Sodium 10000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?