missing translation for 'onlineSavingsMsg'
Learn More

Majantol 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30310368
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310368 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310368 Supplier Dr. Ehrenstorfer Supplier No. DREA14677000AL1000
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Majantol
CAS 103694-68-4
Analyte or Component Names Majantol
Shipping Condition +20°C
Molecular Formula C12 H18 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym β,β,3-Trimethylbenzenepropanol, 2,2-Dimethyl-3-(3-methylphenyl)propan-1-ol, 2,2-Dimethyl-3-(3-methylphenyl)propanol, 2,2-Dimethyl-3-(3-tolyl)propan-1-ol, 2,2-Dimethyl-3-m-tolylpropan-1-ol;
InChI Formula InChI=1S/C12H18O/c1-10-5-4-6-11(7-10)8-12(2,3)9-13/h4-7,13H,8-9H2,1-3H3
SMILES Cc1cccc(CC(C)(C)CO)c1
IUPAC Name 2,2-dimethyl-3-(m-tolyl)propan-1-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 178.27
Quantity 1 mL
Formula Weight 178.1358 g/mol
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.