Learn More
Integra Biosciences™ Mains Adapter
Description
- Mains Adapter for PIPETBOY pro
- Input: 100–240 VAC, 50/60 Hz
- Integrated LED providing an optimal illumination of the area around the pipette for accurate pipetting and preventing eyestrain
- Accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers
- Light as a feather, 190 grams
- Five colors available
- Speed can be set continuously via the thumb wheel
- Red LED indicates when the battery needs to be charged
- Ergonomic design, following the recommendations of hand shape ergonomics
Specifications
Specifications
| Type | Mains Adapter; EU version |
| For Use With (Equipment) | PipetBoy Pro |